Assay Details
Target Gene Details
Entrez Gene ID: | 6229 |
Gene Name: | ribosomal protein S24 |
Gene Aliases: |
DBA3, S24 |
Location: |
Chr.10:78033760-78056813 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPS24 | NM_001142285.1 | 5 | 1599 | NP_001135757.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs79628312] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map