Assay Details
Target Gene Details
Entrez Gene ID: | 57512 |
Gene Name: | G protein-coupled receptor 158 |
Gene Aliases: |
- |
Location: |
Chr.10:25174722-25602229 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPR158 | NM_020752.2 | NP_065803.2 | ||
| AK095120.1 | ||||
| AY528411.1 | AAS18315.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2760160 | Chr.10:25184818 - 25207224 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs114768275] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map