Assay Details
Target Gene Details
Entrez Gene ID: | 57209 |
Gene Name: | zinc finger protein 248 |
Gene Aliases: |
bA162G10.3 |
Location: |
Chr.10:37758452-37858202 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF248 | NM_001267607.1 | NP_001254536.1 | ||
| XM_017016449.1 | XP_016871938.1 | |||
| DA128760.1 |
Target Gene Details
Entrez Gene ID: | 100419868 |
Gene Name: | zinc finger protein 33B pseudogene 1 |
Gene Aliases: |
- |
Location: |
Chr.10:37793938-37795175 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF33BP1 | NR_051997.1 | 1 | 198 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1040704 | Chr.10:37651096 - 38162989 on Build GRCh38 | Gain |
|
| esv3622970 | Chr.10:37789403 - 37797452 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map