Assay Details
Target Gene Details
Entrez Gene ID: | 5791 |
Gene Name: | protein tyrosine phosphatase, receptor type E |
Gene Aliases: |
HPTPE, PTPE, R-PTP-EPSILON |
Location: |
Chr.10:127907061-128085855 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTPRE | NM_001316676.1 | NP_001303605.1 | ||
| NM_001323355.1 | NP_001310284.1 | |||
| NM_001323356.1 | NP_001310285.1 | |||
| NM_006504.5 | NP_006495.1 | |||
| XM_011539994.2 | XP_011538296.1 | |||
| XM_011539995.1 | XP_011538297.1 | |||
| XM_011539996.1 | XP_011538298.1 | |||
| XM_011539998.1 | XP_011538300.1 | |||
| XM_017016467.1 | XP_016871956.1 | |||
| XM_017016469.1 | XP_016871958.1 | |||
| AF406557.1 | AAL01375.1 | |||
| BC050062.1 | AAH50062.1 | |||
| BI559814.1 | ||||
| X54134.1 | CAA38069.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832020 | Chr.10:127816383 - 128009694 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs75304619] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map