Assay Details
Target Gene Details
Entrez Gene ID: | 55853 |
Gene Name: | IDI2 antisense RNA 1 |
Gene Aliases: |
C10orf110, HT009, IDI2-AS |
Location: |
Chr.10:1022637-1044201 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IDI2-AS1 | NR_024628.1 | |||
| NR_024629.1 | ||||
| NR_027708.1 | ||||
| NR_027709.1 | ||||
| AF220183.1 | AAF67648.1 | |||
| AK314921.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2660097 | Chr.10:526001 - 1246069 on Build GRCh38 | Deletion |
|
| nsv5532 | Chr.10:1033559 - 1055760 on Build GRCh38 | Deletion |
|
| nsv1046220 | Chr.10:1018084 - 1143058 on Build GRCh38 | Gain |
|
| esv3891735 | Chr.10:978274 - 1064524 on Build GRCh38 | Gain |
|
| nsv549765 | Chr.10:688289 - 1083587 on Build GRCh38 | Gain |
|
| nsv951231 | Chr.10:1037361 - 1046360 on Build GRCh38 | Duplication |
|
More Information
Additional Information:
For this assay, SNP(s) [rs113814265] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map