Assay Details
Target Gene Details
Entrez Gene ID: | 7417 |
Gene Name: | voltage dependent anion channel 2 |
Gene Aliases: |
POR |
Location: |
Chr.10:75210154-75231448 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VDAC2 | NM_001184783.2 | NP_001171712.1 | ||
| NM_001184823.1 | NP_001171752.1 | |||
| NM_001324087.1 | NP_001311016.1 | |||
| NM_001324088.1 | NP_001311017.1 | |||
| NM_001324089.1 | NP_001311018.1 | |||
| NM_001324090.1 | NP_001311019.1 | |||
| NM_003375.4 | NP_003366.2 | |||
| AK296635.1 | ||||
| AK315919.1 | ||||
| BC000165.2 | AAH00165.2 | |||
| BC012883.1 | AAH12883.2 | |||
| BC072407.1 | AAH72407.1 | |||
| CR456964.1 | CAG33245.1 | |||
| L06328.1 | AAB59457.1 | |||
| L08666.1 | AAA60144.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831913 | Chr.10:75101265 - 75261737 on Build GRCh38 | Loss |
|
| dgv928n100 | Chr.10:75223392 - 75325170 on Build GRCh38 | Gain |
|
| dgv927n100 | Chr.10:75024700 - 75329039 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map