Assay Details
Target Gene Details
Entrez Gene ID: | 120 |
Gene Name: | adducin 3 |
Gene Aliases: |
ADDL, CPSQ3 |
Location: |
Chr.10:109996350-110135565 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ADD3 | NM_001320591.1 | NP_001307520.1 | ||
AK223270.1 | BAD96990.1 | |||
EF553523.1 |
Target Gene Details
Entrez Gene ID: | 100505933 |
Gene Name: | ADD3 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.10:109945559-110008381 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ADD3-AS1 | NR_038943.1 | |||
AI425042.1 | ||||
DB460870.1 | ||||
U92992.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv7547 | Chr.10:109978521 - 110023780 on Build GRCh38 | Deletion |
![]() ![]() |
nsv1052255 | Chr.10:109950120 - 110033444 on Build GRCh38 | Gain |
![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
