Assay Details
Target Gene Details
| Entrez Gene ID: | 23268 | 
| Gene Name: | dynamin binding protein | 
| Gene Aliases: | ARHGEF36, TUBA | 
| Location: | Chr.10:99875571-100009953 on Build GRCh38 | 
| Assay Gene Location: | Within Intron 13 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| DNMBP | NM_001318326.1 | NP_001305255.1 | ||
| NM_001318327.1 | NP_001305256.1 | |||
| NM_015221.3 | NP_056036.1 | |||
| XM_006717735.2 | XP_006717798.1 | |||
| XM_011539559.2 | XP_011537861.1 | |||
| AB023227.1 | BAA76854.1 | |||
| AK128576.1 | ||||
| AK303479.1 | ||||
| AL833283.1 | ||||
| AY196211.1 | AAP34307.1 | |||
| BC041628.1 | AAH41628.1 | 
Target Copy Number Variation Details
| DGV Version: | Release date: 2016-05-15, GRCh GRCh38 | 
| Target Variation | Location | CNV Subtype | Genes | 
|---|---|---|---|
| nsv831960 | Chr.10:99833844 - 99995448 on Build GRCh38 | Gain |  DNMBP-AS1  DNMBP  ABCC2 | 
| nsv552045 | Chr.10:99862766 - 100074335 on Build GRCh38 | Gain |  DNMBP-AS1  DNMBP  CPN1 | 
| nsv1046825 | Chr.10:99861014 - 99912354 on Build GRCh38 | Loss |  DNMBP | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Intronic  Non-exonic  DGV Variation | 
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top