Assay Details
Target Gene Details
Entrez Gene ID: | 8516 |
Gene Name: | integrin subunit alpha 8 |
Gene Aliases: |
- |
Location: |
Chr.10:15513949-15720335 on Build GRCh38 |
Assay Gene Location: | Within Intron 28 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ITGA8 | NM_001291494.1 | NP_001278423.1 | ||
| NM_003638.2 | NP_003629.2 | |||
| AK297740.1 | ||||
| L36531.1 | AAA93514.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv679n100 | Chr.10:15529099 - 15664910 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map