Assay Details
Target Gene Details
Entrez Gene ID: | 26019 |
Gene Name: | UPF2 regulator of nonsense transcripts homolog (yeast) |
Gene Aliases: |
HUPF2, RENT2, smg-3 |
Location: |
Chr.10:11920022-12043170 on Build GRCh38 |
Assay Gene Location: | Within Intron 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UPF2 | NM_015542.3 | NP_056357.1 | ||
| NM_080599.2 | NP_542166.1 | |||
| XM_017016034.1 | XP_016871523.1 | |||
| AB037829.1 | ||||
| AF301013.1 | AAG33225.1 | |||
| AF318574.1 | AAG60689.1 | |||
| AK000764.1 | BAA91369.1 | |||
| AK074375.1 | BAB85064.1 | |||
| AK096191.1 | BAC04721.1 | |||
| AK124930.1 | 1 | 498 | ||
| AK295209.1 | ||||
| AL080198.1 | CAB45771.1 | |||
| AY013249.1 | AAG48509.1 | |||
| BC114964.1 | ||||
| BC115737.1 | ||||
| BX647325.1 | 1 | 423 |
More Information
Additional Information:
For this assay, SNP(s) [rs112166002,rs72235385] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map