Assay Details
Target Gene Details
Entrez Gene ID: | 220972 |
Gene Name: | membrane associated ring-CH-type finger 8 |
Gene Aliases: |
CMIR, MARCH-VIII, MIR, RNF178, c-MIR |
Location: |
Chr.10:45454585-45594907 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MARCH8 | NM_001002266.2 | NP_001002266.1 | ||
| NM_001282866.1 | NP_001269795.1 | |||
| NM_145021.5 | NP_659458.2 | |||
| XM_005271804.2 | XP_005271861.1 | |||
| XM_006717704.2 | XP_006717767.1 | |||
| XM_011539492.2 | XP_011537794.1 | |||
| XM_011539493.2 | XP_011537795.1 | |||
| XM_011539494.2 | XP_011537796.1 | |||
| XM_011539495.1 | XP_011537797.1 | |||
| XM_017015894.1 | XP_016871383.1 | |||
| AK057289.1 | ||||
| AK307667.1 | ||||
| AK313340.1 | ||||
| AL833316.1 | ||||
| BC025394.2 | AAH25394.1 | |||
| BC066988.1 | AAH66988.1 | |||
| BG433948.1 | ||||
| BG717635.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1043536 | Chr.10:45420493 - 45580787 on Build GRCh38 | Gain |
|
| esv3891797 | Chr.10:45008026 - 45674466 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map