Assay Details
Target Gene Details
Entrez Gene ID: | 79746 |
Gene Name: | enoyl-CoA hydratase domain containing 3 |
Gene Aliases: |
- |
Location: |
Chr.10:11742357-11764070 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ECHDC3 | NM_024693.4 | 1 | 123 | NP_078969.2 |
| XM_011519689.1 | 1 | 98 | XP_011517991.1 | |
| AF275677.1 | 1 | 97 | AAG24387.1 | |
| AF289604.1 | 1 | 79 | AAL55788.1 | |
| AK024562.1 | 1 | 36 | BAB14928.1 | |
| AK222511.1 | 1 | 62 | BAD96231.1 | |
| AK290902.1 | 1 | 53 | ||
| BC009617.1 | 1 | 45 | AAH09617.1 | |
| BP332406.1 | 1 | 123 |
More Information
Additional Information:
For this assay, SNP(s) [rs76272655] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map