Assay Details
Target Gene Details
Entrez Gene ID: | 56288 |
Gene Name: | par-3 family cell polarity regulator |
Gene Aliases: |
ASIP, Baz, PAR3, PAR3alpha, PARD-3, PARD3A, PPP1R118, SE2-5L16, SE2-5LT1, SE2-5T2 |
Location: |
Chr.10:34109560-34815325 on Build GRCh38 |
Assay Gene Location: | Within Intron 25 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831830 | Chr.10:34022936 - 34158791 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map