Assay Details
Target Gene Details
Entrez Gene ID: | 56952 |
Gene Name: | phosphoribosyl transferase domain containing 1 |
Gene Aliases: |
HHGP |
Location: |
Chr.10:24848607-24952644 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PRTFDC1 | NM_001282786.1 | NP_001269715.1 | ||
NM_020200.6 | NP_064585.1 | |||
AB209792.1 | BAD93029.1 | |||
AF226056.1 | AAF86956.1 | |||
AK021950.1 | BAB13944.1 | |||
AK222674.1 | BAD96394.1 | |||
AK294130.1 | ||||
AK304055.1 | ||||
AK309614.1 | ||||
BC008662.2 | AAH08662.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
