Assay Details
Target Gene Details
Entrez Gene ID: | 9748 |
Gene Name: | STE20 like kinase |
Gene Aliases: |
LOSK, STK2, bA16H23.1, se20-9 |
Location: |
Chr.10:103967173-104029233 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLK | NM_001304743.1 | NP_001291672.1 | ||
| NM_014720.3 | NP_055535.2 | |||
| XM_011540401.2 | XP_011538703.1 | |||
| AB002804.1 | BAA19655.1 | |||
| AF273048.1 | AAG34908.1 | |||
| AK307389.1 | ||||
| BC047762.1 | AAH47762.1 | |||
| BC047885.1 | AAH47885.1 | |||
| BC064804.1 | AAH64804.1 | |||
| BC111565.1 | AAI11566.1 | |||
| D86959.1 | BAA13195.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv951008 | Chr.10:103973043 - 103983942 on Build GRCh38 | Duplication |
|
| dgv967n100 | Chr.10:103975777 - 104014012 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map