Assay Details
Target Gene Details
Entrez Gene ID: | 54518 |
Gene Name: | amyloid beta precursor protein binding family B member 1 interacting protein |
Gene Aliases: |
INAG1, PREL1, RARP1, RIAM |
Location: |
Chr.10:26438324-26567879 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| APBB1IP | NM_019043.3 | NP_061916.3 | ||
| XM_011519514.2 | XP_011517816.1 | |||
| AK000812.1 | BAA91384.1 | |||
| AK303597.1 | ||||
| AK308916.1 | ||||
| AY152730.1 | AAN75525.1 | |||
| BC035636.1 | AAH35636.1 | |||
| BC054516.1 | AAH54516.1 | |||
| BM150270.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1043832 | Chr.10:26380111 - 26520471 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map