Assay Details
Target Gene Details
Entrez Gene ID: | 953 |
Gene Name: | ectonucleoside triphosphate diphosphohydrolase 1 |
Gene Aliases: |
ATPDase, CD39, NTPDase-1, SPG64 |
Location: |
Chr.10:95710901-95877266 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Target Gene Details
Entrez Gene ID: | 728558 |
Gene Name: | ENTPD1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.10:95753206-96090235 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ENTPD1-AS1 | NR_038444.1 | |||
| AK091396.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv950997 | Chr.10:95762744 - 95809443 on Build GRCh38 | Duplication |
|
| nsv1052213 | Chr.10:95777038 - 95802184 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map