Assay Details
Target Gene Details
Entrez Gene ID: | 114815 |
Gene Name: | sortilin related VPS10 domain containing receptor 1 |
Gene Aliases: |
hSorCS |
Location: |
Chr.10:106573663-107181138 on Build GRCh38 |
Assay Gene Location: | Within Intron 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SORCS1 | NM_001013031.2 | NP_001013049.1 | ||
| NM_001206569.1 | NP_001193498.1 | |||
| NM_001206570.1 | NP_001193499.1 | |||
| NM_001206571.1 | NP_001193500.1 | |||
| NM_001206572.1 | NP_001193501.1 | |||
| NM_052918.4 | NP_443150.3 | |||
| XM_011539199.2 | XP_011537501.1 | |||
| XM_017015614.1 | XP_016871103.1 | |||
| XM_017015615.1 | XP_016871104.1 | |||
| XM_017015616.1 | XP_016871105.1 | |||
| XM_017015617.1 | XP_016871106.1 | |||
| XM_017015618.1 | XP_016871107.1 | |||
| AB209142.1 | BAD92379.1 | |||
| AF284756.1 | AAL56667.1 | |||
| AK074581.1 | ||||
| AK123752.1 | ||||
| AK125464.1 | ||||
| AK126905.1 | ||||
| AK289797.1 | ||||
| AY099452.1 | AAM43811.1 | |||
| AY099453.1 | AAM43812.1 | |||
| BC131597.1 | ||||
| EU281343.1 | ||||
| EU281344.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831977 | Chr.10:106466255 - 106653420 on Build GRCh38 | Gain |
|
| nsv1047446 | Chr.10:105797401 - 106616713 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs145113017] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map