Assay Details
Target Gene Details
Entrez Gene ID: | 23268 |
Gene Name: | dynamin binding protein |
Gene Aliases: |
ARHGEF36, TUBA |
Location: |
Chr.10:99875571-100009953 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DNMBP | NM_001318326.1 | NP_001305255.1 | ||
NM_001318327.1 | NP_001305256.1 | |||
NM_015221.3 | NP_056036.1 | |||
XM_006717735.2 | XP_006717798.1 | |||
XM_011539559.2 | XP_011537861.1 | |||
AB023227.1 | BAA76854.1 | |||
AK128576.1 | ||||
AK303479.1 | ||||
AL833283.1 | ||||
AY196211.1 | AAP34307.1 | |||
BC041628.1 | AAH41628.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831960 | Chr.10:99833844 - 99995448 on Build GRCh38 | Gain |
![]() ![]() ![]() |
nsv552045 | Chr.10:99862766 - 100074335 on Build GRCh38 | Gain |
![]() ![]() ![]() |
nsv1046825 | Chr.10:99861014 - 99912354 on Build GRCh38 | Loss |
![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
