Assay Details
Target Gene Details
Entrez Gene ID: | 63877 |
Gene Name: | family with sequence similarity 204 member A |
Gene Aliases: |
C10orf84, bA319I23.1 |
Location: |
Chr.10:118305889-118342352 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM204A | NM_001134672.1 | NP_001128144.1 | ||
| NM_022063.2 | NP_071346.1 | |||
| XM_005270024.1 | XP_005270081.1 | |||
| AK023250.1 | BAB14487.1 | |||
| AK026630.1 | BAB15514.1 | |||
| BC023577.2 | AAH23577.1 | |||
| BM837313.1 | ||||
| CR457304.1 | CAG33585.1 |

Genomic Map