Assay Details
Target Gene Details
Entrez Gene ID: | 953 |
Gene Name: | ectonucleoside triphosphate diphosphohydrolase 1 |
Gene Aliases: |
ATPDase, CD39, NTPDase-1, SPG64 |
Location: |
Chr.10:95710901-95877266 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ENTPD1 | NM_001098175.1 | NP_001091645.1 | ||
| XM_011540371.2 | XP_011538673.1 | |||
| AJ133133.1 | CAB41886.1 | |||
| AJ133134.1 | CAB41887.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv526180 | Chr.10:95572438 - 95751280 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map