Assay Details
Target Gene Details
Entrez Gene ID: | 100507470 |
Gene Name: | GRID1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.10:85577731-85607215 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GRID1-AS1 | NR_038986.1 | |||
| AK097655.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv7476 | Chr.10:85540344 - 85585215 on Build GRCh38 | Deletion |
|
| esv3891878 | Chr.10:85560815 - 85628886 on Build GRCh38 | Loss |
|
| nsv1052427 | Chr.10:85234813 - 86270793 on Build GRCh38 | Gain |
|

Genomic Map