Assay Details
Target Gene Details
Entrez Gene ID: | 10133 |
Gene Name: | optineurin |
Gene Aliases: |
ALS12, FIP2, GLC1E, HIP7, HYPL, NRP, TFIIIA-INTP |
Location: |
Chr.10:13100082-13138276 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OPTN | NM_001008211.1 | NP_001008212.1 | ||
| NM_001008212.1 | NP_001008213.1 | |||
| NM_001008213.1 | NP_001008214.1 | |||
| NM_021980.4 | NP_068815.2 | |||
| AF049614.1 | AAC26850.1 | |||
| AF061034.1 | AAC16046.1 | |||
| AF420371.1 | AAL76327.1 | |||
| AF420372.1 | AAL76328.1 | |||
| AF420373.1 | AAL76329.1 | |||
| AK055403.1 | ||||
| AK122746.1 | ||||
| AK303715.1 | ||||
| AK316171.1 | ||||
| BC013876.2 | AAH13876.1 | |||
| BC032762.1 | AAH32762.1 | |||
| KU178502.1 | ||||
| KU178503.1 | ||||
| KU178504.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2762845 | Chr.10:13087065 - 13142762 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map