Assay Details
Target Gene Details
Entrez Gene ID: | 387700 |
Gene Name: | solute carrier family 16 member 12 |
Gene Aliases: |
CJMG, CRT2, MCT12 |
Location: |
Chr.10:89430294-89536028 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC16A12 | NM_213606.3 | NP_998771.3 | ||
| XM_017016237.1 | XP_016871726.1 | |||
| XM_017016238.1 | XP_016871727.1 | |||
| XM_017016239.1 | XP_016871728.1 | |||
| AK124901.1 | BAC85987.1 | |||
| AK127303.1 | BAC86925.1 | |||
| BC086873.1 | AAH86873.1 | |||
| CR936657.1 |
More Information
Set Membership: |
|

Genomic Map