Assay Details
Target Gene Details
Entrez Gene ID: | 119391 |
Gene Name: | glutathione S-transferase omega 2 |
Gene Aliases: |
GSTO 2-2, bA127L20.1 |
Location: |
Chr.10:104267626-104304948 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GSTO2 | XM_011539270.2 | 1 | 481 | XP_011537572.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831971 | Chr.10:104120100 - 104301309 on Build GRCh38 | Gain |
|
| nsv552062 | Chr.10:104254126 - 104282744 on Build GRCh38 | Loss |
|
| nsv1039166 | Chr.10:104260183 - 104302423 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map