Assay Details
Target Gene Details
Entrez Gene ID: | 10105 |
Gene Name: | peptidylprolyl isomerase F |
Gene Aliases: |
CYP3, CyP-M, Cyp-D, CypD |
Location: |
Chr.10:79347464-79355334 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPIF | NM_005729.3 | NP_005720.1 | ||
| XM_005269379.2 | XP_005269436.1 | |||
| AK296669.1 | ||||
| AK312757.1 | ||||
| BC005020.1 | AAH05020.1 | |||
| BC110299.1 | AAI10300.1 | |||
| BE791220.1 | ||||
| M80254.1 | AAA58434.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1161981 | Chr.10:79309656 - 79385886 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map