Assay Details
Target Gene Details
Entrez Gene ID: | 55506 |
Gene Name: | H2A histone family member Y2 |
Gene Aliases: |
macroH2A2 |
Location: |
Chr.10:70052601-70112284 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
H2AFY2 | NM_018649.2 | NP_061119.1 | ||
AF151534.1 | AAF72101.1 | |||
AF336304.1 | AAK52471.1 | |||
AK022475.1 | BAB14049.1 | |||
AK022776.1 | BAB14239.1 | |||
AK131057.1 | ||||
BC016172.2 | AAH16172.1 | |||
CX752382.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv32737 | Chr.10:69504230 - 70786847 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv551479 | Chr.10:70007598 - 70179618 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
