Assay Details
Target Gene Details
Entrez Gene ID: | 8027 |
Gene Name: | signal transducing adaptor molecule |
Gene Aliases: |
STAM-1, STAM1 |
Location: |
Chr.10:17644125-17716824 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STAM | NM_001324282.1 | NP_001311211.1 | ||
| NM_001324283.1 | NP_001311212.1 | |||
| NM_001324284.1 | NP_001311213.1 | |||
| NM_001324285.1 | NP_001311214.1 | |||
| NM_001324286.1 | NP_001311215.1 | |||
| NM_001324287.1 | NP_001311216.1 | |||
| NM_001324288.1 | NP_001311217.1 | |||
| NM_001324289.1 | NP_001311218.1 | |||
| NM_003473.3 | NP_003464.1 | |||
| XM_011519695.2 | XP_011517997.1 | |||
| AK303083.1 | ||||
| AK314405.1 | ||||
| AU118708.1 | ||||
| BC030586.2 | AAH30586.1 | |||
| U43899.1 | AAC50734.1 |
More Information
Set Membership: |
|

Genomic Map