Assay Details
Target Gene Details
Entrez Gene ID: | 1791 |
Gene Name: | DNA nucleotidylexotransferase |
Gene Aliases: |
TDT |
Location: |
Chr.10:96304328-96338564 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNTT | NM_001017520.1 | NP_001017520.1 | ||
| NM_004088.3 | NP_004079.3 | |||
| AB046378.1 | BAB72001.1 | |||
| AK223317.1 | BAD97037.1 | |||
| AK313615.1 | ||||
| BC012920.1 | AAH12920.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2761620 | Chr.10:96287288 - 96329992 on Build GRCh38 | Gain |
|
| nsv831949 | Chr.10:96167838 - 96385945 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs78039628] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map