Assay Details
Target Gene Details
Entrez Gene ID: | 83759 |
Gene Name: | RNA binding motif protein 4B |
Gene Aliases: |
RBM30, RBM4L, ZCCHC15, ZCCHC21B, ZCRB3B |
Location: |
Chr.11:66664994-66677921 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RBM4B | NM_001286135.1 | NP_001273064.1 | ||
| NM_031492.3 | NP_113680.1 | |||
| XM_011545297.2 | XP_011543599.1 | |||
| XM_017018402.1 | XP_016873891.1 | |||
| AK095158.1 | ||||
| BC003503.1 | AAH03503.1 | |||
| BC004951.1 | AAH04951.1 | |||
| BQ428264.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv518919 | Chr.11:66641938 - 66700998 on Build GRCh38 | Loss |
|
| nsv522277 | Chr.11:66466552 - 66726354 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map