Assay Details
Target Gene Details
Entrez Gene ID: | 100526737 |
Gene Name: | RBM14-RBM4 readthrough |
Gene Aliases: |
COAZ |
Location: |
Chr.11:66616582-66646473 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RBM14-RBM4 | NM_001198845.1 | NP_001185774.1 | ||
| NM_001198846.1 | NP_001185775.1 | |||
| AW958943.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522277 | Chr.11:66466552 - 66726354 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs77658314] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map