Assay Details
Target Gene Details
Entrez Gene ID: | 84188 |
Gene Name: | fatty acyl-CoA reductase 1 |
Gene Aliases: |
MLSTD2, PFCRD, SDR10E1 |
Location: |
Chr.11:13668659-13732346 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAR1 | NM_032228.5 | NP_115604.1 | ||
| XM_011520400.1 | XP_011518702.1 | |||
| AK026381.1 | BAB15467.1 | |||
| AK315527.1 | ||||
| AY358784.1 | AAQ89144.1 | |||
| AY423606.1 | AAR84086.1 | |||
| AY600449.1 | AAT42129.1 | |||
| BC017377.2 | AAH17377.1 | |||
| CR936619.1 | CAI56762.1 | |||
| DA515509.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs371845778] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map