Assay Details
Target Gene Details
Entrez Gene ID: | 9973 |
Gene Name: | copper chaperone for superoxide dismutase |
Gene Aliases: |
- |
Location: |
Chr.11:66593162-66606019 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCS | NM_005125.1 | NP_005116.1 | ||
| AF002210.1 | AAC51764.1 | |||
| AY113179.1 | AAM50090.1 | |||
| BC105016.1 | AAI05017.1 | |||
| BC112055.1 | AAI12056.1 | |||
| CR541928.1 | CAG46726.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522277 | Chr.11:66466552 - 66726354 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map