Assay Details
Target Gene Details
Entrez Gene ID: | 4162 |
Gene Name: | melanoma cell adhesion molecule |
Gene Aliases: |
CD146, MUC18 |
Location: |
Chr.11:119308524-119317132 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MCAM | NM_006500.2 | NP_006491.2 | ||
XM_017017759.1 | XP_016873248.1 | |||
XM_017017760.1 | XP_016873249.1 | |||
XM_017017761.1 | XP_016873250.1 | |||
XM_017017762.1 | XP_016873251.1 | |||
AB209925.1 | BAD93162.1 | |||
AF089868.1 | AAD17799.1 | |||
AK128335.1 | ||||
AK291571.1 | ||||
AK312430.1 | ||||
BC056418.1 | AAH56418.1 | |||
M28882.1 | AAA20922.1 | |||
M29277.1 | AAA20824.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv952747 | Chr.11:119308591 - 119317490 on Build GRCh38 | Deletion |
![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
