Assay Details
Target Gene Details
| Entrez Gene ID: | 4684 | 
| Gene Name: | neural cell adhesion molecule 1 | 
| Gene Aliases: | CD56, MSK39, NCAM | 
| Location: | Chr.11:112961247-113278436 on Build GRCh38 | 
| Assay Gene Location: | Within Intron 1 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| NCAM1 | NM_000615.6 | NP_000606.3 | ||
| NM_001076682.3 | NP_001070150.1 | |||
| NM_001242607.1 | NP_001229536.1 | |||
| NM_001242608.1 | NP_001229537.1 | |||
| NM_181351.4 | NP_851996.2 | |||
| AB209443.1 | BAD92680.1 | |||
| AF005070.1 | AAB61615.1 | |||
| AK292453.1 | ||||
| AK314589.1 | ||||
| BC029119.1 | ||||
| BC047244.1 | AAH47244.1 | |||
| DA231540.1 | ||||
| DA337649.1 | ||||
| S71824.1 | AAB31836.1 | |||
| U63041.1 | AAB04558.1 | |||
| X16841.1 | CAA34739.1 | 
Target Copy Number Variation Details
| DGV Version: | Release date: 2016-05-15, GRCh GRCh38 | 
| Target Variation | Location | CNV Subtype | Genes | 
|---|---|---|---|
| nsv832271 | Chr.11:112972081 - 113007925 on Build GRCh38 | Gain |  NCAM1 | 
| dgv1279n100 | Chr.11:112966837 - 112987859 on Build GRCh38 | Loss |  NCAM1 | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Intronic  Non-exonic  DGV Variation | 
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top