Assay Details
Target Gene Details
Entrez Gene ID: | 2113 |
Gene Name: | ETS proto-oncogene 1, transcription factor |
Gene Aliases: |
ETS-1, EWSR2, c-ets-1, p54 |
Location: |
Chr.11:128458761-128587593 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ETS1 | NM_001143820.1 | NP_001137292.1 | ||
| NM_001162422.1 | NP_001155894.1 | |||
| NM_005238.3 | NP_005229.1 | |||
| XM_011542649.2 | XP_011540951.1 | |||
| XM_011542650.1 | XP_011540952.1 | |||
| XM_011542651.1 | XP_011540953.1 | |||
| XM_011542652.1 | XP_011540954.1 | |||
| XM_017017314.1 | XP_016872803.1 | |||
| XM_017017315.1 | XP_016872804.1 | |||
| XM_017017316.1 | XP_016872805.1 | |||
| XM_017017317.1 | XP_016872806.1 | |||
| AK291840.1 | ||||
| AK301405.1 | ||||
| AY943926.1 | ||||
| BC017314.2 | AAH17314.1 | |||
| BT019452.1 | AAV38259.1 | |||
| BT019453.1 | AAV38260.1 | |||
| BX640634.1 | CAE45783.1 | |||
| CR542254.1 | CAG47050.1 | |||
| J04101.1 | AAA52410.1 | |||
| X14798.1 | CAA32904.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv442246 | Chr.11:128463624 - 128465155 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map