Assay Details
Target Gene Details
Entrez Gene ID: | 322 |
Gene Name: | amyloid beta precursor protein binding family B member 1 |
Gene Aliases: |
FE65, MGC:9072, RIR |
Location: |
Chr.11:6395124-6419830 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| APBB1 | NM_001164.4 | NP_001155.1 | ||
| NM_145689.2 | NP_663722.1 | |||
| NR_047512.2 | ||||
| XM_006718209.3 | XP_006718272.2 | |||
| XM_006718210.3 | XP_006718273.2 | |||
| XM_006718211.1 | XP_006718274.1 | |||
| XM_006718212.3 | XP_006718275.2 | |||
| XM_006718213.1 | XP_006718276.1 | |||
| XM_006718214.3 | XP_006718277.2 | |||
| XM_006718215.3 | XP_006718278.3 | |||
| XM_011520038.2 | XP_011518340.1 | |||
| XM_011520040.2 | XP_011518342.1 | |||
| XM_017017639.1 | XP_016873128.1 | |||
| XM_017017640.1 | XP_016873129.1 | |||
| XM_017017641.1 | XP_016873130.1 | |||
| AF394214.1 | AAL79526.1 | |||
| AK293452.1 | ||||
| AK293613.1 | ||||
| AK294495.1 | ||||
| AK297544.1 | ||||
| AK311725.1 | ||||
| BC010854.2 | AAH10854.1 | |||
| L77864.1 | AAB93631.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv553371 | Chr.11:6374460 - 6413235 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map