Assay Details
Target Gene Details
Entrez Gene ID: | 472 |
Gene Name: | ATM serine/threonine kinase |
Gene Aliases: |
AT1, ATA, ATC, ATD, ATDC, ATE, TEL1, TELO1 |
Location: |
Chr.11:108222484-108369102 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATM | NM_000051.3 | NP_000042.3 | ||
| XM_005271561.4 | XP_005271618.2 | |||
| XM_005271562.4 | XP_005271619.2 | |||
| XM_006718843.3 | XP_006718906.1 | |||
| XM_011542840.2 | XP_011541142.1 | |||
| XM_011542842.2 | XP_011541144.1 | |||
| XM_011542843.2 | XP_011541145.1 | |||
| XM_011542844.2 | XP_011541146.1 | |||
| XM_017017789.1 | XP_016873278.1 | |||
| XM_017017790.1 | XP_016873279.1 | |||
| XM_017017791.1 | XP_016873280.1 | |||
| XM_017017792.1 | XP_016873281.1 | |||
| KF704396.1 | ||||
| U33841.1 | AAC50289.1 | |||
| X91196.1 | CAA62603.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv556430 | Chr.11:108226606 - 108304735 on Build GRCh38 | Loss |
|
| dgv2181n54 | Chr.11:108227415 - 108244860 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map