Assay Details
Target Gene Details
Entrez Gene ID: | 9177 |
Gene Name: | 5-hydroxytryptamine receptor 3B |
Gene Aliases: |
5-HT3B |
Location: |
Chr.11:113904796-113949682 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HTR3B | NM_006028.4 | NP_006019.1 | ||
| XM_011543063.1 | XP_011541365.1 | |||
| AF080582.1 | AAD12242.1 | |||
| AF169255.1 | AAF03691.1 | |||
| AK314268.1 | ||||
| AM293589.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832273 | Chr.11:113784031 - 113957584 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs78249783] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map