Assay Details
Target Gene Details
Entrez Gene ID: | 2313 |
Gene Name: | Fli-1 proto-oncogene, ETS transcription factor |
Gene Aliases: |
EWSR2, SIC-1 |
Location: |
Chr.11:128685263-128813267 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FLI1 | NM_001271012.1 | NP_001257941.1 | ||
| XM_011542701.2 | XP_011541003.1 | |||
| XM_017017405.1 | XP_016872894.1 | |||
| XM_017017406.1 | XP_016872895.1 | |||
| AK294279.1 |
Target Gene Details
Entrez Gene ID: | 100507392 |
Gene Name: | smooth muscle and endothelial cell enriched migration/differentiation-associated long non-coding RNA |
Gene Aliases: |
FLI1-AS1, lncRNA9 |
Location: |
Chr.11:128691672-128696023 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SENCR | NR_038908.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv826128 | Chr.11:128689312 - 128695562 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map