Assay Details
Target Gene Details
Entrez Gene ID: | 341116 |
Gene Name: | membrane spanning 4-domains A10 |
Gene Aliases: |
CD20L7, MS4A9 |
Location: |
Chr.11:60785332-60801305 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MS4A10 | NM_206893.3 | NP_996776.2 | ||
| XM_011544989.1 | XP_011543291.1 | |||
| XM_011544991.2 | XP_011543293.1 | |||
| AK122633.1 | BAC85498.1 | |||
| BC137259.1 | ||||
| BC137263.1 |
More Information
Set Membership: |
|

Genomic Map