Assay Details
Target Gene Details
Entrez Gene ID: | 60529 |
Gene Name: | ALX homeobox 4 |
Gene Aliases: |
CRS5, FND2 |
Location: |
Chr.11:44260728-44310166 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ALX4 | NM_021926.3 | NP_068745.2 | ||
| AB058691.1 | ||||
| AF294629.1 | AAG23961.1 | |||
| AJ404888.1 | CAC15060.1 | |||
| AK092346.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2761670 | Chr.11:44279998 - 44284831 on Build GRCh38 | Loss |
|
| esv3626160 | Chr.11:44279795 - 44284852 on Build GRCh38 | Loss |
|
| esv3547679 | Chr.11:44279785 - 44284950 on Build GRCh38 | Deletion |
|
| dgv344e212 | Chr.11:44278235 - 44285381 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map