Assay Details
Target Gene Details
Entrez Gene ID: | 84649 |
Gene Name: | diacylglycerol O-acyltransferase 2 |
Gene Aliases: |
ARAT, GS1999FULL, HMFN1045 |
Location: |
Chr.11:75768733-75801536 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DGAT2 | NM_001253891.1 | NP_001240820.1 | ||
| NM_032564.4 | NP_115953.2 | |||
| XM_011545304.2 | XP_011543606.1 | |||
| AB048286.2 | BAB40641.2 | |||
| AF384161.2 | AAK84176.2 | |||
| AL834287.1 | CAD38961.1 | |||
| AY358532.1 | AAQ88896.1 | |||
| AY780646.1 | ||||
| AY780647.1 | AAV35727.1 | |||
| BC015234.1 | AAH15234.1 | |||
| DB462004.1 |
More Information
Set Membership: |
|

Genomic Map