Assay Details
Target Gene Details
Entrez Gene ID: | 91801 |
Gene Name: | alkB homolog 8, tRNA methyltransferase |
Gene Aliases: |
ABH8, TRM9, TRMT9 |
Location: |
Chr.11:107502726-107565740 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ALKBH8 | NM_001301010.1 | NP_001287939.1 | ||
| NM_138775.2 | NP_620130.2 | |||
| XM_005271728.2 | XP_005271785.1 | |||
| XM_005271729.3 | XP_005271786.1 | |||
| XM_017018553.1 | XP_016874042.1 | |||
| XM_017018554.1 | XP_016874043.1 | |||
| XM_017018555.1 | XP_016874044.1 | |||
| XM_017018556.1 | XP_016874045.1 | |||
| AB218768.1 | ||||
| AK293603.1 | ||||
| AK298800.1 | ||||
| AK304413.1 | ||||
| BC015183.1 | AAH15183.1 | |||
| BX649085.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv556376 | Chr.11:107380125 - 107615872 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map