Assay Details
Target Gene Details
Entrez Gene ID: | 283104 |
Gene Name: | SBF2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.11:9758293-9811319 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SBF2-AS1 | NR_036485.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2744095 | Chr.11:9623376 - 10133785 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() |
nsv1045550 | Chr.11:9696777 - 9768541 on Build GRCh38 | Gain |
![]() ![]() ![]() |
nsv553468 | Chr.11:9646281 - 9806324 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
nsv1053369 | Chr.11:9626290 - 9854507 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |
