Assay Details
Target Gene Details
Entrez Gene ID: | 322 |
Gene Name: | amyloid beta precursor protein binding family B member 1 |
Gene Aliases: |
FE65, MGC:9072, RIR |
Location: |
Chr.11:6395124-6419830 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv553371 | Chr.11:6374460 - 6413235 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map