Assay Details
Target Gene Details
Entrez Gene ID: | 10313 |
Gene Name: | reticulon 3 |
Gene Aliases: |
ASYIP, HAP, NSPL2, NSPLII, RTN3-A1 |
Location: |
Chr.11:63681315-63759891 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832183 | Chr.11:63649206 - 63797093 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map