Assay Details
Target Gene Details
Entrez Gene ID: | 100526771 |
Gene Name: | TMPRSS4 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.11:118015772-118086793 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMPRSS4-AS1 | NR_038318.1 | |||
| AB231754.1 | ||||
| AB231755.1 | ||||
| AB231756.1 | ||||
| AB231757.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3579922 | Chr.11:118039084 - 118047247 on Build GRCh38 | Loss |
|
| esv998313 | Chr.11:118046416 - 118050916 on Build GRCh38 | Insertion |
|
| nsv509444 | Chr.11:118033438 - 118117536 on Build GRCh38 | Insertion |
|

Genomic Map