Assay Details
Target Gene Details
Entrez Gene ID: | 55107 |
Gene Name: | anoctamin 1 |
Gene Aliases: |
DOG1, ORAOV2, TAOS2, TMEM16A |
Location: |
Chr.11:70078302-70189545 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv555328 | Chr.11:70098564 - 70132159 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map