Assay Details
Target Gene Details
Entrez Gene ID: | 4684 |
Gene Name: | neural cell adhesion molecule 1 |
Gene Aliases: |
CD56, MSK39, NCAM |
Location: |
Chr.11:112961247-113278436 on Build GRCh38 |
Assay Gene Location: | Within Intron 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NCAM1 | NM_000615.6 | NP_000606.3 | ||
| NM_001242607.1 | NP_001229536.1 | |||
| NM_181351.4 | NP_851996.2 | |||
| AK057509.1 | ||||
| AK292453.1 | ||||
| AK299865.1 | ||||
| AK314589.1 | ||||
| BC014205.2 | AAH14205.2 | |||
| BC047244.1 | AAH47244.1 | |||
| M17410.1 | AAA59913.1 | |||
| S71824.1 | AAB31836.1 | |||
| U63041.1 | AAB04558.1 | |||
| X55322.1 |
Target Gene Details
Entrez Gene ID: | 100288346 |
Gene Name: | NCAM1 antisense RNA1 |
Gene Aliases: |
- |
Location: |
Chr.11:113269532-113273901 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NCAM1-AS1 | NR_034101.1 | |||
| AK293718.1 | ||||
| DC305174.1 | ||||
| DC309426.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv483080 | Chr.11:113216072 - 113400474 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map